2-Bromo-2'-chloro-5'-(trifluoromethyl)acetophenone
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Chloro-5-(trifluoromethyl)phenacyl bromide
PC10425-1G
PC10425-5G
APOSPC10425-1GEA
6620
CZK
APOSPC10425-1G
APOSPC10425-5G
2-Bromo-2'-chloro-5'-(trifluoromethyl)acetophenone
2-Bromo-2'-chloro-5'-(trifluoromethyl)acetophenone
Vzorec:
C₉H₅BrClF₃O Bod tání: 27…29 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03094501 CAS číslo: 79779-64-9 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...