3'-Amino-4'-fluoroacetanilide
Dodavatel: Apollo Scientific
|
Varování
|
PC10414-1G
PC10414-250MG
APOSPC10414-1GEA
2900
CZK
APOSPC10414-1G
APOSPC10414-250MG
3'-Amino-4'-fluoroacetanilide
3'-Amino-4'-fluoroacetanilide
Vzorec:
C₈H₉FN₂O Storage Temperature: Ambient |
MDL Number:
MFCD07368688 CAS číslo: 113596-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...