2-Bromo-3',4',5'-trifluoroacetophenone
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3,4,5-Trifluorophenacyl bromide
PC10407-1G
PC10407-5G
APOSPC10407-1GEA
5460
CZK
APOSPC10407-1G
APOSPC10407-5G
2-Bromo-3',4',5'-trifluoroacetophenone
2-Bromo-3',4',5'-trifluoroacetophenone
Vzorec:
C₈H₄BrF₃O Storage Temperature: Refrigerator |
MDL Number:
MFCD07368686 CAS číslo: 443914-94-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...