Tisk…

3,5-Bis(trifluoromethyl)benzenesulphonamide

Dodavatel: Apollo Scientific
Varování

Synonyms: 3,5-Bis(trifluoromethyl)benzene sulphonamide

PC10406-1G PC10406-5G
APOSPC10406-1GEA 1660 CZK
APOSPC10406-1G APOSPC10406-5G
3,5-Bis(trifluoromethyl)benzenesulphonamide
3,5-Bis(trifluoromethyl)benzenesulphonamide
Vzorec: C₈H₅F₆NO₂S
Storage Temperature: Ambient
MDL Number: MFCD01935138
CAS číslo: 39213-22-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR