Tisk…

α,α,α-Trifluoro-p-toluidine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Aminobenzotrifluoride, 4-(Trifluoromethyl)aniline, Leflunomide impurity A

PC1040-1KG PC1040-250G
APOSPC1040-1KGEA 4960 CZK
APOSPC1040-1KG APOSPC1040-250G
α,α,α-Trifluoro-p-toluidine 97%
α,α,α-Trifluoro-p-toluidine

Precursor to 2-(4-aminophenyl)-substituted tetrahydropyrimidines & imidazolines

Vzorec: C₇H₆F₃N
MW: 161.13
Bod varu: 118 °C/60mmHg
Bod tání: 7…9 °C
Hustota: 1.3
Bod vzplanutí: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00064396
CAS číslo: 455-14-1
EINECS: 207-236-5
UN: 2810
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR