α-Bromo-2,3,5,6-tetrafluorotoluene
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2,3,5,6-Tetrafluorobenzyl bromide, 2,3,5,6-Tetrafluorobenzylbromide
PC10395-1G
PC10395-5G
APOSPC10395-1GEA
2070
CZK
APOSPC10395-1G
APOSPC10395-5G
α-Bromo-2,3,5,6-tetrafluorotoluene
α-Bromo-2,3,5,6-tetrafluorotoluene
Vzorec:
C₇H₃BrF₄ MW: 243 g/mol Bod varu: 79…80 °C/80mmHg Hustota: 1,79 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013549 CAS číslo: 53001-73-3 EINECS: 258-289-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...