2-(2-Pyridinyl)-4-(trifluoromethyl)-5-pyrimidinecarboxylic acid
Dodavatel: Apollo Scientific
PC10374-1G
PC10374-500MG
APOSPC10374-1GEA
15180
CZK
APOSPC10374-1G
APOSPC10374-500MG
2-(2-Pyridinyl)-4-(trifluoromethyl)-5-pyrimidinecarboxylic acid
2-(2-Pyridinyl)-4-(trifluoromethyl)-5-pyrimidinecarboxylic acid
Vzorec:
C₁₁H₆F₃N₃O₂ MW: 269,18 g/mol Bod tání: 224…226 °C Storage Temperature: Ambient |
MDL Number:
MFCD06200983 CAS číslo: 874816-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...