Methyl-2-amino-5-(3-fluorophenyl)-1,3-thiazole-4-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
PC10369-10G
PC10369-1G
PC10369-5G
APOSPC10369-10GEA
15540
CZK
APOSPC10369-10G
APOSPC10369-1G
APOSPC10369-5G
Methyl-2-amino-5-(3-fluorophenyl)-1,3-thiazole-4-carboxylate
Methyl-2-amino-5-(3-fluorophenyl)-1,3-thiazole-4-carboxylate
Vzorec:
C₁₁H₉FN₂O₂S MW: 252,27 g/mol Bod tání: 232…234 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06200922 CAS číslo: 127918-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...