Tisk…

1,4-Difluoro-2-methyl-5-nitrobenzene

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,5-Difluoro-4-nitrotoluene

PC10366-1G PC10366-25G PC10366-5G PC10366-100G
APOSPC10366-1GEA 417 CZK
APOSPC10366-1G APOSPC10366-25G APOSPC10366-5G APOSPC10366-100G
1,4-Difluoro-2-methyl-5-nitrobenzene
1,4-Difluoro-2-methyl-5-nitrobenzene
Vzorec: C₇H₅F₂NO₂
MW: 173,1169 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06200905
CAS číslo: 141412-60-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR