Tisk…

6-Fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 1,4-Dihydro-6-fluoro-4-oxoquinoline-3-carboxylic acid

PC10350-1G PC10350-5G
APOSPC10350-1GEA 2070 CZK
APOSPC10350-1G APOSPC10350-5G
6-Fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid
6-Fluoro-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid
Vzorec: C₁₀H₆FNO₃
MW: 207,16 g/mol
Bod tání: 298…300 °C
Storage Temperature: Ambient
MDL Number: MFCD03425825
CAS číslo: 117685-48-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR