Tisk…

3-Chloro-5-fluorobenzylamine 97%

Dodavatel: Apollo Scientific

PC1035-5G PC1035-1G
APOSPC1035-5GEA 4300 CZK
APOSPC1035-5G APOSPC1035-1G
3-Chloro-5-fluorobenzylamine 97%
3-Chloro-5-fluorobenzylamine
Vzorec: C₇H₇ClFN
MW: 159,59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06213787
CAS číslo: 90390-33-3
UN: 1760
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR