Pentafluorophenyl-4-methylbenzenesulphonate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Pentafluorophenyl toluene-4-sulphonate
PC10315-1G
PC10315-5G
APOSPC10315-1GEA
1330
CZK
APOSPC10315-1G
APOSPC10315-5G
Pentafluorophenyl-4-methylbenzenesulphonate
Pentafluorophenyl 4-methylbenzenesulphonate
Vzorec:
C₁₃H₇F₅O₃S Bod tání: 65…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060056 CAS číslo: 2069-36-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...