Pentafluorophenyl-3-nitrobenzenesulphonate
Dodavatel: Apollo Scientific
PC10313-1G
PC10313-500MG
PC10313-5G
APOSPC10313-1GEA
7840
CZK
APOSPC10313-1G
APOSPC10313-500MG
APOSPC10313-5G
Pentafluorophenyl-3-nitrobenzenesulphonate
Pentafluorophenyl-3-nitrobenzenesulphonate
Vzorec:
C₁₂H₄F₅NO₅S Bod tání: 67…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD05975121 CAS číslo: 663175-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...