Tisk…

Pentafluorophenyl-4-ethylbenzenesulphonate

Dodavatel: Apollo Scientific

PC10312-1G PC10312-500MG PC10312-5G
APOSPC10312-1GEA 7840 CZK
APOSPC10312-1G APOSPC10312-500MG APOSPC10312-5G
Pentafluorophenyl-4-ethylbenzenesulphonate
Pentafluorophenyl-4-ethylbenzenesulphonate
Vzorec: C₁₄H₉F₅O₃S
Bod tání: 61…63 °C
Storage Temperature: Ambient
MDL Number: MFCD05975134
CAS číslo: 885950-41-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR