Tisk…

(E)-3-[2-(3,5-Difluorophenoxy)phenyl]-2-propenoic acid

Dodavatel: Apollo Scientific

PC10295-1G PC10295-500MG PC10295-5G
APOSPC10295-1GEA 15180 CZK
APOSPC10295-1G APOSPC10295-500MG APOSPC10295-5G
(E)-3-[2-(3,5-Difluorophenoxy)phenyl]-2-propenoic acid
(E)-3-[2-(3,5-Difluorophenoxy)phenyl]-2-propenoic acid
Vzorec: C₁₅H₁₀F₂O₃
Bod tání: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD05663813
CAS číslo: 886361-03-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR