4-(3,5-Difluorophenoxy)benzaldehyde
Dodavatel: Apollo Scientific
PC10294-1G
PC10294-5G
APOSPC10294-1GEA
12370
CZK
APOSPC10294-1G
APOSPC10294-5G
4-(3,5-Difluorophenoxy)benzaldehyde
4-(3,5-Difluorophenoxy)benzaldehyde
Vzorec:
C₁₃H₈F₂O₂ Bod tání: 57…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD05663812 CAS číslo: 886361-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...