Tisk…

3-(1,3-Benzoxazol-2-yl)-4-fluoroaniline

Dodavatel: Apollo Scientific

PC10293-1G PC10293-500MG
APOSPC10293-1GEA 15180 CZK
APOSPC10293-1G APOSPC10293-500MG
3-(1,3-Benzoxazol-2-yl)-4-fluoroaniline
3-(1,3-Benzoxazol-2-yl)-4-fluoroaniline
Vzorec: C₁₃H₉FN₂O
Bod tání: 153…156 °C
Storage Temperature: Ambient
MDL Number: MFCD03425815
CAS číslo: 220705-28-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR