1,1,1-Trifluoro-3-[4-(2-fluorophenyl)piperazino]-2-propanol
Dodavatel: Apollo Scientific
PC10289-1G
PC10289-500MG
APOSPC10289-1GEA
26320
CZK
APOSPC10289-1G
APOSPC10289-500MG
1,1,1-Trifluoro-3-[4-(2-fluorophenyl)piperazino]-2-propanol
1,1,1-Trifluoro-3-[4-(2-fluorophenyl)piperazino]-2-propanol
Vzorec:
C₁₃H₁₆F₄N₂O Bod tání: 92…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD01871727 CAS číslo: 453557-80-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...