Tisk…

1,1,1-Trifluoro-3-{[2-(trifluoromethyl)benzyl]amino}-2-propanol

Dodavatel: Apollo Scientific

PC10288-1G PC10288-500MG
APOSPC10288-1GEA 22750 CZK
APOSPC10288-1G APOSPC10288-500MG
1,1,1-Trifluoro-3-{[2-(trifluoromethyl)benzyl]amino}-2-propanol
1,1,1-Trifluoro-3-{[2-(trifluoromethyl)benzyl]amino}-2-propanol
Vzorec: C₁₁H₁₁F₆NO
MW: 287,21 g/mol
Bod tání: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD01871716
CAS číslo: 453557-79-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR