Tisk…

5-Chloro-6-oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Chloro-1,6-dihydro-6-oxo-1-[3-(trifluoromethyl)benzyl]pyridine-3-carboxylic acid

PC10286-1G PC10286-500MG
APOSPC10286-1GEA 11810 CZK
APOSPC10286-1G APOSPC10286-500MG
5-Chloro-6-oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid
5-Chloro-6-oxo-1-[3-(trifluoromethyl)benzyl]-1,6-dihydro-3-pyridine carboxylic acid
Vzorec: C₁₄H₉ClF₃NO₃
MW: 331,68 g/mol
Bod tání: 191…193 °C
Storage Temperature: Ambient
MDL Number: MFCD00140108
CAS číslo: 339024-06-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR