Tisk…

4-Chloro-2-fluoro-5-[(4-fluorobenzyl)oxy]aniline

Dodavatel: Apollo Scientific

PC10281-1G PC10281-500MG PC10281-5G
APOSPC10281-1GEA 15180 CZK
APOSPC10281-1G APOSPC10281-500MG APOSPC10281-5G
4-Chloro-2-fluoro-5-[(4-fluorobenzyl)oxy]aniline
4-Chloro-2-fluoro-5-[(4-fluorobenzyl)oxy]aniline
Vzorec: C₁₃H₁₀ClF₂NO
Bod tání: 87…89 °C
Storage Temperature: Ambient
MDL Number: MFCD03425812
CAS číslo: 453557-77-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR