1-(2-Fluoro-4-nitrophenyl)piperazine
Dodavatel: Apollo Scientific
|
Varování
|
PC10278-1G
PC10278-25G
PC10278-5G
APOSPC10278-1GEA
4170
CZK
APOSPC10278-1G
APOSPC10278-25G
APOSPC10278-5G
1-(2-Fluoro-4-nitrophenyl)piperazine
1-(2-Fluoro-4-nitrophenyl)piperazine
Vzorec:
C₁₀H₁₂FN₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01572463 CAS číslo: 154590-33-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...