Methyl-1-(3-fluorobenzyl)-4-piperidinecarboxylate
Dodavatel: Apollo Scientific
PC10273-1G
PC10273-500MG
PC10273-5G
APOSPC10273-1GEA
6190
CZK
APOSPC10273-1G
APOSPC10273-500MG
APOSPC10273-5G
Methyl-1-(3-fluorobenzyl)-4-piperidinecarboxylate
Methyl-1-(3-fluorobenzyl)-4-piperidinecarboxylate
Vzorec:
C₁₄H₁₈FNO₂ MW: 251.3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03425805 CAS číslo: 383146-94-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...