Tisk…

2,2,2-Trichloro-1-[4-(4-fluorobenzoyl)-1H-pyrrol-2-yl]-1-ethanone

Dodavatel: Apollo Scientific

Synonyms: 4-(4-Fluorobenzoyl)-2-(trichloroacetyl)-1H-pyrrole

PC10272-1G PC10272-500MG
APOSPC10272-1GEA 11540 CZK
APOSPC10272-1G APOSPC10272-500MG
2,2,2-Trichloro-1-[4-(4-fluorobenzoyl)-1H-pyrrol-2-yl]-1-ethanone
2,2,2-Trichloro-1-[4-(4-fluorobenzoyl)-1H-pyrrol-2-yl]-1-ethanone
Vzorec: C₁₃H₇Cl₃FNO₂
MW: 334,56 g/mol
Bod tání: 202…204 °C
Storage Temperature: Ambient
MDL Number: MFCD03001281
CAS číslo: 453557-67-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR