Tisk…

(4-Fluorophenoxy)acetonitrile

Dodavatel: Apollo Scientific
Varování

PC10268-10G PC10268-5G
APOSPC10268-10GEA 7210 CZK
APOSPC10268-10G APOSPC10268-5G
(4-Fluorophenoxy)acetonitrile
(4-Fluorophenoxy)acetonitrile
Vzorec: C₈H₆FNO
Storage Temperature: Ambient
CAS číslo: 24115-20-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR