Tisk…

1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-ol

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-Hydroxy-1-methyl-3-(trifluoromethyl)-1H-pyrazole

PC10250-5G PC10250-10G
APOSPC10250-5GEA 3060 CZK
APOSPC10250-5G APOSPC10250-10G
1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-ol
1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-ol
Vzorec: C₅H₅F₃N₂O
MW: 166,1 g/mol
Bod tání: 178…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00139833
CAS číslo: 122431-37-2
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR