Tisk…

Methyl-4-{3-amino-3-oxo-1-[(2,2,2-trifluoroacetyl)amino]propyl}benzenecarboxylate

Dodavatel: Apollo Scientific

PC10223-1G PC10223-500MG PC10223-5G
APOSPC10223-1GEA 15180 CZK
APOSPC10223-1G APOSPC10223-500MG APOSPC10223-5G
Methyl-4-{3-amino-3-oxo-1-[(2,2,2-trifluoroacetyl)amino]propyl}benzenecarboxylate
Methyl-4-{3-amino-3-oxo-1-[(2,2,2-trifluoroacetyl)amino]propyl}benzenecarboxylate
Vzorec: C₁₃H₁₃F₃N₂O₄
MW: 318,25 g/mol
Bod tání: 231…233 °C
Storage Temperature: Ambient
MDL Number: MFCD03791206
CAS číslo: 886360-57-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR