Tisk…

1-(3-Fluorobenzyl)-4-piperidinecarboxylic acid hydrochloride

Dodavatel: Apollo Scientific

PC10210-1G PC10210-500MG PC10210-5G
APOSPC10210-1GEA 6190 CZK
APOSPC10210-1G APOSPC10210-500MG APOSPC10210-5G
1-(3-Fluorobenzyl)-4-piperidinecarboxylic acid hydrochloride
1-(3-Fluorobenzyl)-4-piperidinecarboxylic acid hydrochloride
Vzorec: C₁₃H₁₇ClFNO₂
MW: 273,74 g/mol
Bod tání: 220…223 °C
Storage Temperature: Ambient
MDL Number: MFCD03425740
CAS číslo: 451485-55-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR