Tisk…

{4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanol

Dodavatel: Apollo Scientific

PC10199-1G PC10199-2.5G
APOSPC10199-1GEA 8200 CZK
APOSPC10199-1G APOSPC10199-2.5G
{4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanol
{4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methanol
Vzorec: C₁₂H₁₀F₃NOS
Bod tání: 120…123 °C
Storage Temperature: Ambient
MDL Number: MFCD03791170
CAS číslo: 317318-96-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR