Tisk…

[1-Methyl-5-(4-methylphenoxy)-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol

Dodavatel: Apollo Scientific

PC10195-1G PC10195-500MG PC10195-5G
APOSPC10195-1GEA 15180 CZK
APOSPC10195-1G APOSPC10195-500MG APOSPC10195-5G
[1-Methyl-5-(4-methylphenoxy)-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
[1-Methyl-5-(4-methylphenoxy)-3-(trifluoromethyl)-1H-pyrazol-4-yl]methanol
Vzorec: C₁₃H₁₃F₃N₂O₂
Bod tání: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD03001207
CAS číslo: 400073-97-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR