Tisk…

3-{4-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}acrylic acid

Dodavatel: Apollo Scientific

PC10190-1G PC10190-500MG PC10190-5G
APOSPC10190-1GEA 7840 CZK
APOSPC10190-1G APOSPC10190-500MG APOSPC10190-5G
3-{4-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}acrylic acid
3-{4-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}acrylic acid
Vzorec: C₁₆H₁₂ClFO₃
Bod tání: 177…179 °C
Storage Temperature: Ambient
MDL Number: MFCD02091011
CAS číslo: 400825-72-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR