3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenol
Dodavatel: Apollo Scientific
PC10179-500MG
PC10179-1G
APOSPC10179-500MGEA
4370
CZK
APOSPC10179-500MG
APOSPC10179-1G
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenol
3-{[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy}benzenol
Vzorec:
C₁₂H₇ClF₃NO₂ Bod tání: 100…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00172750 CAS číslo: 95711-07-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...