Tisk…

N'-Hydroxy-2-[3-(trifluoromethyl)phenoxy]ethanimidamide

Dodavatel: Apollo Scientific

PC10176-1G PC10176-5G
APOSPC10176-1GEA 5200 CZK
APOSPC10176-1G APOSPC10176-5G
N'-Hydroxy-2-[3-(trifluoromethyl)phenoxy]ethanimidamide
N'-Hydroxy-2-[3-(trifluoromethyl)phenoxy]ethanimidamide
Vzorec: C₉H₉F₃N₂O₂
MW: 234,18 g/mol
Bod tání: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD02090466
CAS číslo: 690632-15-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR