Tisk…

Methyl-2-amino-5-(2-fluorophenyl)-1,3-thiazole-4-carboxylate

Dodavatel: Apollo Scientific

PC10171-10G PC10171-1G PC10171-5G
APOSPC10171-10GEA 7210 CZK
APOSPC10171-10G APOSPC10171-1G APOSPC10171-5G
Methyl-2-amino-5-(2-fluorophenyl)-1,3-thiazole-4-carboxylate
Methyl-2-amino-5-(2-fluorophenyl)-1,3-thiazole-4-carboxylate
Vzorec: C₁₁H₉FN₂O₂S
MW: 252,27 g/mol
Bod tání: 203 °C
Storage Temperature: Ambient
MDL Number: MFCD06200924
CAS číslo: 886361-38-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR