Tisk…

{3-[3-(Trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}methanol

Dodavatel: Apollo Scientific

PC10170-1G PC10170-500MG PC10170-5G
APOSPC10170-1GEA 15180 CZK
APOSPC10170-1G APOSPC10170-500MG APOSPC10170-5G
{3-[3-(Trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}methanol
{3-[3-(Trifluoromethyl)phenyl]-1,2,4-oxadiazol-5-yl}methanol
Vzorec: C₁₀H₇F₃N₂O₂
Bod tání: 66…67 °C
Storage Temperature: Ambient
MDL Number: MFCD06200920
CAS číslo: 886361-32-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR