4-[(2-Fluorobenzyl)oxy]-N'-hydroxybenzenecarboximidamide
Dodavatel: Apollo Scientific
PC10168-1G
PC10168-500MG
APOSPC10168-1GEA
13460
CZK
APOSPC10168-1G
APOSPC10168-500MG
4-[(2-Fluorobenzyl)oxy]-N'-hydroxybenzenecarboximidamide
4-[(2-Fluorobenzyl)oxy]-N'-hydroxybenzenecarboximidamide
Vzorec:
C₁₄H₁₃FN₂O₂ Bod tání: 138…139 °C Storage Temperature: Ambient |
MDL Number:
MFCD00662630 CAS číslo: 261965-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...