Tisk…

2-[(3-Fluorobenzyl)sulphanyl]benzenecarboxylic acid

Dodavatel: Apollo Scientific

Synonyms: 2-(3-Fluorobenzylthio)benzoic acid

PC10145-1G PC10145-500MG PC10145-5G
APOSPC10145-1GEA 4900 CZK
APOSPC10145-1G APOSPC10145-500MG APOSPC10145-5G
2-[(3-Fluorobenzyl)sulphanyl]benzenecarboxylic acid
2-[(3-Fluorobenzyl)sulphanyl]benzenecarboxylic acid
Vzorec: C₁₄H₁₁FO₂S
MW: 262,3 g/mol
Bod tání: 183…186 °C
Storage Temperature: Ambient
MDL Number: MFCD02937499
CAS číslo: 385383-44-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR