3-(2,6-Dimethylmorpholino)-1,1,1-trifluoro-2-propanol
Dodavatel: Apollo Scientific
PC10140-1G
PC10140-250MG
APOSPC10140-1GEA
13690
CZK
APOSPC10140-1G
APOSPC10140-250MG
3-(2,6-Dimethylmorpholino)-1,1,1-trifluoro-2-propanol
3-(2,6-Dimethylmorpholino)-1,1,1-trifluoro-2-propanol
Vzorec:
C₉H₁₆F₃NO₂ MW: 227,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03001226 CAS číslo: 667938-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...