2,5-Difluoro-4-nitrobenzoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
PC10116-1G
PC10116-25G
PC10116-5G
APOSPC10116-1GEA
622
CZK
APOSPC10116-1G
APOSPC10116-25G
APOSPC10116-5G
2,5-Difluoro-4-nitrobenzoic acid 98%
2,5-Difluoro-4-nitrobenzoic acid
Vzorec:
C₇H₃F₂NO₄ MW: 203,10 g/mol Bod tání: 145…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD03425705 CAS číslo: 116465-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...