2-(4-Methylphenyl)-1-hydrazinecarbothioamide
Dodavatel: Apollo Scientific
Synonyms:
Methyl 4-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}benzenecarboxylate
PC10112-1G
PC10112-500MG
PC10112-5G
APOSPC10112-1GEA
4900
CZK
APOSPC10112-1G
APOSPC10112-500MG
APOSPC10112-5G
2-(4-Methylphenyl)-1-hydrazinecarbothioamide
2-(4-Methylphenyl)-1-hydrazinecarbothioamide
Vzorec:
C₈H₁₁N₃S MW: 181,26 g/mol Bod tání: 79…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD03791137 CAS číslo: 7382-40-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...