1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylic acid
Dodavatel: Apollo Scientific
PC10111-1G
PC10111-500MG
APOSPC10111-1GEA
6950
CZK
APOSPC10111-1G
APOSPC10111-500MG
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylic acid
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylic acid
Vzorec:
C₁₂H₁₂ClF₃N₂O₂ Bod tání: 135…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00172245 CAS číslo: 337919-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...