Ethyl 3,5-bis(trifluoromethyl)phenylacetate 97%
Dodavatel: Apollo Scientific
Synonyms:
Ethyl 2-(3,5-bis(trifluoromethyl)phenyl)acetate
PC1011-25G
PC1011-50G
PC1011-5G
APOSPC1011-25GEA
15740
CZK
APOSPC1011-25G
APOSPC1011-50G
APOSPC1011-5G
Ethyl 3,5-bis(trifluoromethyl)phenylacetate 97%
Ethyl-3,5-bis(trifluoromethyl)phenylacetate
Vzorec:
C₁₂H₁₀F₆O₂ MW: 300.20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08689894 CAS číslo: 144632-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...