Tisk…

2-Fluoro-3-methoxyphenylboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Fluoro-3-methoxybenzeneboronic acid, (2-Fluoro-3-methoxyphenyl)boronic acid

PC10096-100G PC10096-25G PC10096-5G
APOSPC10096-100GEA 9100 CZK
APOSPC10096-100G APOSPC10096-25G APOSPC10096-5G
2-Fluoro-3-methoxyphenylboronic acid
2-Fluoro-3-methoxyphenylboronic acid
Vzorec: C₇H₈BFO₃
MW: 169,95 g/mol
Bod tání: 117…122 °C
Storage Temperature: Ambient
MDL Number: MFCD02094728
CAS číslo: 352303-67-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR