1,2,4-Trichloro-1,1,2-trifluorobutane
Dodavatel: Apollo Scientific
|
Varování
|
PC10073-1G
PC10073-5G
PC10073-10G
APOSPC10073-1GEA
1660
CZK
APOSPC10073-1G
APOSPC10073-5G
APOSPC10073-10G
1,2,4-Trichloro-1,1,2-trifluorobutane
1,2,4-Trichloro-1,1,2-trifluorobutane
Vzorec:
C₄H₄Cl₃F₃ Bod varu: 134…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00077597 CAS číslo: 261760-97-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...