Tisk…

2-Nitrotetrafluoroaniline

Dodavatel: Apollo Scientific
Varování

PC0998-100MG PC0998-250MG
APOSPC0998-100MGEA 827 CZK
APOSPC0998-100MG APOSPC0998-250MG
2-Nitrotetrafluoroaniline
2-Nitrotetrafluoroaniline
Vzorec: C₆H₂F₄N₂O₂
MW: 210,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00278791
CAS číslo: 6157-98-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR