Tisk…

3-Nitro-4-(trifluoromethyl)phenol 98%

Dodavatel: Apollo Scientific

PC0997-1G PC0997-5G PC0997-10G
APOSPC0997-1GEA 1370 CZK
APOSPC0997-1G APOSPC0997-5G APOSPC0997-10G
3-Nitro-4-(trifluoromethyl)phenol 98%
3-Nitro-4-(trifluoromethyl)phenol
Vzorec: C7H4F3NO3
Bod tání: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD07783660
CAS číslo: 25889-36-5
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR