Tisk…

1,1,1,3,3-Pentachloro-3-fluoroacetone 95%

Dodavatel: Apollo Scientific

Synonyms: Pentachlorofluoroacetone

PC0978-25G PC0978-5G PC0978-1G
APOSPC0978-25GEA 16730 CZK
APOSPC0978-25G APOSPC0978-5G APOSPC0978-1G
1,1,1,3,3-Pentachloro-3-fluoroacetone 95%
1,1,1,3,3-Pentachloro-3-fluoroacetone
Vzorec: C₃Cl₅FO
MW: 248,29 g/mol
Bod varu: 162…164 °C
Bod vzplanutí: >100 °C
Storage Temperature: Ambient
CAS číslo: 2378-08-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR