2-(Pentafluorophenoxy)ethanol 95%
Dodavatel: Apollo Scientific
Synonyms:
2-(Perfluorophenoxy)ethanol
PC0975-25G
PC0975-5G
PC0975-1G
APOSPC0975-25GEA
15540
CZK
APOSPC0975-25G
APOSPC0975-5G
APOSPC0975-1G
2-(Pentafluorophenoxy)ethanol 95%
2-(Pentafluorophenoxy)ethanol
Vzorec:
C₈H₅F₅O₂ MW: 228,12 g/mol Bod varu: 78…80 °C/4mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00191478 CAS číslo: 2192-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...