Tisk…

α-Chloro-2,6-difluorotoulene 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-(Chloromethyl)-1,3-difluorobenzene, 2,6-Difluorobenzyl chloride, 1-(Chloromethyl)-2,6-difluorobenzene

PC0971-1G PC0971-100G
APOSPC0971-1GEA 417 CZK
APOSPC0971-1G APOSPC0971-100G
α-Chloro-2,6-difluorotoulene 98%
α-Chloro-2,6-difluorotoulene
Vzorec: C₇H₅ClF₂
MW: 162.57
Bod varu: 172…173 °C/640mmHg
Bod tání: 32…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00191346
CAS číslo: 697-73-4
UN: 1759
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR