2-Fluoro-4-(methylsulphonyl)aniline 99%
Dodavatel: Apollo Scientific
|
Varování
|
PC0964-25G
PC0964-5G
PC0964-100G
APOSPC0964-25GEA
4700
CZK
APOSPC0964-25G
APOSPC0964-5G
APOSPC0964-100G
2-Fluoro-4-(methylsulphonyl)aniline 99%
2-Fluoro-4-(methylsulphonyl)aniline
Vzorec:
C₇H₈FNO₂S MW: 189,21 g/mol Bod tání: 145…147 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06800756 CAS číslo: 832755-13-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...